Synonym: Sodium hydrogen sulfite
CAS Number 7631-90-5
Linear Formula NaHSO3
Molecular Weight 104.06
MDL number MFCD00003530
PubChem Substance ID 24848149
| Related Categories | Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents |
| grade | purum |
| assay | ~40% |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?