EMAIL THIS PAGE TO A FRIEND
13530 Sigma-Aldrich

Benzylmalonic acid

purum, ≥98.0% (T)

Synonym: β-Phenylisosuccinic acid

  • CAS Number 616-75-1

  • Linear Formula C6H5CH2CH(COOH)2

  • Molecular Weight 194.18

  •  Beilstein Registry Number 643530

  •  EC Number 210-491-5

  •  MDL number MFCD00004261

  •  PubChem Substance ID 329750207

Purchase

Properties

Related Categories Building Blocks, C10, Carbonyl Compounds, Carboxylic Acid Monomers, Carboxylic Acids,
InChI Key   JAEJSNFTJMYIEF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   117-120 °C(lit.)
  117-120 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
OO0525000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

100773 Benzylmalonic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?