13571 Sigma-Aldrich

Sodium sulfate decahydrate

puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.0% (calc. to the dried substance)

DOWNLOAD MSDS (PDF)

Synonym: Glauber’s salt

  • CAS Number 7727-73-3

  • Linear Formula Na2SO4 · 10H2O

  • Molecular Weight 322.20

  •  EC Number 231-820-9

  •  MDL number MFCD00149189

  •  PubChem Substance ID 24848217

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Sodium, Synthetic Reagents
grade   puriss.
assay   ≥99.0% (calc. to the dried substance)
pharmacopeia   testing & handling conforms to Pharmacopeia
quality   meets analytical specification of Ph. Eur., BP, USP
impurities   acidity or alkalinity, complies
  residual solvents, complies
  ≤10 mg/kg heavy metals (as Pb)
  ≤200 mg/kg free alkali (as NaOH)
  ≤300 mg/kg free acid (as H2SO4)
loss   53-57% loss on drying, 130 °C
density   1.46 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤100 mg/kg
cation traces   As: ≤2 mg/kg
  Ca: ≤50 mg/kg
  Fe: ≤5 mg/kg
  Mg: ≤100 mg/kg
  Zn: ≤20 mg/kg
suitability   complies for appearance of solution
  complies for identity

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Sodium sulfate decahydrate

ACS reagent, ≥99.0%

Sodium sulfate decahydrate

reagent grade, 97%

Sodium sulfate decahydrate

BioUltra, ≥99.0% (calc. on dry substance, T)

Sodium sulfate decahydrate

puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?