• USA Home
  • 13610 - Benzyl 2-nitrophenyl ether

EMAIL THIS PAGE TO A FRIEND
13610 Sigma-Aldrich

Benzyl 2-nitrophenyl ether

Selectophore, ≥99.0%

  • CAS Number 4560-41-2

  • Empirical Formula (Hill Notation) C13H11NO3

  • Molecular Weight 229.23

  •  Beilstein Registry Number 1287342

  •  MDL number MFCD00143680

  •  PubChem Substance ID 329750041

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Plasticizer for ISE More...
grade   Selectophore
InChI Key   ZYWSXGRMDPBISP-UHFFFAOYSA-N
assay   ≥99.0% (GC)
  ≥99.0%
refractive index   n20/D 1.598
density   1.225 g/mL at 20 °C(lit.)

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer in neutral carrier based polymer membrane ion-selective electrodes

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?