EMAIL THIS PAGE TO A FRIEND
13635 Sigma-Aldrich

1-Z-4-Piperidone

purum, ≥95.0% (HPLC)

Synonym: 1-(Benzyloxycarbonyl)-4-piperidinone, 1-Cbz-4-Piperidone, Benzyl 4-oxo-1-piperidinecarboxylate

  • CAS Number 19099-93-5

  • Empirical Formula (Hill Notation) C13H15NO3

  • Molecular Weight 233.26

  •  Beilstein Registry Number 1533716

  •  MDL number MFCD00673144

  •  PubChem Substance ID 329750051

Purchase

Properties

Related Categories Building Blocks, C13, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   VZOVOHRDLOYBJX-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (HPLC)
refractive index   n20/D 1.542(lit.)
  n20/D 1.542
bp   114-140 °C/0.25 mmHg(lit.)
density   1.172 g/mL at 25 °C(lit.)

Description

General description

1-Z-4-Piperidone is the starting material in biheteroaromatic diphosphine oxides-catalyzed stereoselective direct aldol condensation reactions.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?