13803 Sigma-Aldrich

Sulfur

99.5-100.5%, meets analytical specification of Ph. Eur., BP, USP, puriss., precipitated

DOWNLOAD MSDS (PDF)
  • CAS Number 7704-34-9

  • Empirical Formula (Hill Notation) S

  • Molecular Weight 32.07

  •  EC Number 231-722-6

  •  MDL number MFCD00085316

  •  PubChem Substance ID 24848229

Properties

Related Categories Acaricides, Analytical Standards, Analytical/Chromatography, Chromatography, Environmental Standards,
vapor density   8.9 (vs air)
vapor pressure   1 mmHg ( 183.8 °C)
  10 mmHg ( 246 °C)
grade   puriss.
assay   99.5-100.5%
autoignition temp.   450 °F
quality   meets analytical specification of Ph. Eur., BP, USP
  precipitated
resistivity   2E23 μΩ-cm20°C
impurities   acidity or alkalinity, complies
  residual solvents, complies
  ≤0.01% free alkali (as NaOH)
  ≤0.02% free acid (as H2SO4)
  ≤0.5% water (Karl Fischer)
ign. residue   ≤0.2% (as SO4)
  ≤0.3%
bp   444.7 °C(lit.)
mp   112.8 °C (rhombic)(lit.)
  117-120 °C(lit.)
  119.0 °C (monoclinic)(lit.)
solubility   carbon disulfide: in accordance1 g/5mL
anion traces   chloride (Cl-): ≤100 mg/kg
  sulfate (SO42-): ≤100 mg/kg
  sulfide (S2-): ≤10 mg/kg
cation traces   As: ≤40 mg/kg
suitability   complies for appearance of solution
  corresponds for odor

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Sulfur
36576Sulfur

PESTANAL®, analytical standard

Sulfur
84683Sulfur

purum p.a., ≥99.5% (T)

Sulfur
13825Sulfur

powder, colloidal

Sulfur
84684Sulfur

Ph Eur

Sulfur
213292Sulfur

99.998% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?