13825 Sigma-Aldrich

Sulfur

powder, colloidal

DOWNLOAD MSDS (PDF)
  • CAS Number 7704-34-9

  • Empirical Formula (Hill Notation) S

  • Molecular Weight 32.07

  •  EC Number 231-722-6

  •  MDL number MFCD00085316

  •  PubChem Substance ID 24848240

Properties

vapor density   8.9 (vs air)
vapor pressure   1 mmHg ( 183.8 °C)
  10 mmHg ( 246 °C)
form   colloidal
  powder
autoignition temp.   450 °F
resistivity   2E23 μΩ-cm20°C
bp   444.7 °C(lit.)
mp   112.8 °C (rhombic)(lit.)
  117-120 °C(lit.)
  119.0 °C (monoclinic)(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Sulfur
36576Sulfur

PESTANAL®, analytical standard

Sulfur
84683Sulfur

purum p.a., ≥99.5% (T)

Sulfur
13803Sulfur

99.5-100.5%, meets analytical specification of Ph. Eur., BP, USP, puriss., precipitated

Sulfur
84684Sulfur

Ph Eur

Sulfur
414980Sulfur

powder, 99.98% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?