• USA Home
  • 13947 - Benzyltriethylammonium bromide

EMAIL THIS PAGE TO A FRIEND
13947 Sigma-Aldrich

Benzyltriethylammonium bromide

purum, ≥98.0% (AT)

  • CAS Number 5197-95-5

  • Linear Formula C6H5CH2N(Br)(C2H5)3

  • Molecular Weight 272.22

  •  Beilstein Registry Number 3739113

  •  EC Number 225-986-1

  •  MDL number MFCD00011822

  •  PubChem Substance ID 329750375

Purchase

Properties

InChI Key   CHQVQXZFZHACQQ-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (AT)
mp   193-195 °C (dec.)(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

147125 Benzyltriethylammonium bromide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?