• USA Home
  • 13970 - Benzyltrimethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
13970 Sigma-Aldrich

Benzyltrimethylammonium chloride

purum, ≥98.0% (AT)

  • CAS Number 56-93-9

  • Linear Formula C6H5CH2N(Cl)(CH3)3

  • Molecular Weight 185.69

  •  Beilstein Registry Number 3917255

  •  EC Number 200-300-3

  •  MDL number MFCD00011782

  •  PubChem Substance ID 24848426

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   KXHPPCXNWTUNSB-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (AT)
mp   230-235 °C (dec.)
  239 °C (dec.)(lit.)

Description

Other Notes

This product has been replaced by 228982-ALDRICH | Benzyltrimethylammonium chloride 97%

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
2
RTECS 
BO8400000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?