• USA Home
  • 14002 - Benzyltrimethylammonium tribromide

EMAIL THIS PAGE TO A FRIEND
14002 Sigma-Aldrich

Benzyltrimethylammonium tribromide

purum, ≥97.0% (AT)

Synonym: BTMABr3, Benzyltrimethylammonium bromide dibromide

  • CAS Number 111865-47-5

  • Linear Formula C6H5CH2N(Br3)(CH3)3

  • Molecular Weight 389.95

  •  Beilstein Registry Number 7105839

  •  MDL number MFCD00134423

Purchase

Properties

Related Categories Ammonium, Ammonium Salts, Bromination, C-X Bond Formation (Halogen), Chemical Synthesis,
InChI Key   OQESKQAHRXOSMS-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   99-101 °C(lit.)
  99-101 °C

Description

Other Notes

Convenient-to-handle, solid brominating reagent with a greater brominating potential than tetrabutylammonium tribromide. BTMABr3 has been used for the bromination of phenols, methyl ketones, and aromatic amines ; Hofmann degradation of amides; Reagent for oxidations

Same product — same quality — now Sigma-Aldrich brand

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

376426 Benzyltrimethylammonium tribromide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?