Synonym: Titania, Titanium dioxide
CAS Number 13463-67-7
Linear Formula TiO2
Molecular Weight 79.87
EC Number 236-675-5
MDL number MFCD00011269
PubChem Substance ID 24848462
| Related Categories | Materials Science, Metal and Ceramic Science, Oxides, Titanium |
| grade | puriss. |
| assay | 98.0-100.5% |
| 99-100.5% | |
| 99.0-100.5% (calc. on dry substance) | |
| quality | meets analytical specification of Ph. Eur., BP, USP |
| impurities | acidity or alkalinity, complies |
| residual solvents, complies |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?