EMAIL THIS PAGE TO A FRIEND
14089 Sigma-Aldrich

Succinic anhydride

puriss., ≥99.0% (NT)

Synonym: Dihydro-2,5-furandione

  • CAS Number 108-30-5

  • Empirical Formula (Hill Notation) C4H4O3

  • Molecular Weight 100.07

  •  Beilstein Registry Number 108441

  •  EC Number 203-570-0

  •  MDL number MFCD00005525

  •  PubChem Substance ID 329750395

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Anhydrides, Chemical Synthesis, Organic Building Blocks More...
vapor density   3.5 (vs air)
vapor pressure   1 mmHg ( 92 °C)
InChI Key   RINCXYDBBGOEEQ-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
bp   261 °C(lit.)
mp   118-120 °C(lit.)
  118-120 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
1
RTECS 
WN0875000
Flash Point(F) 
314.6 °F
Flash Point(C) 
157 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

239690 Succinic anhydride, ≥99% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?