EMAIL THIS PAGE TO A FRIEND
14150 Sigma-Aldrich

Dimethyl succinate

purum, ≥98.0% (GC)

Synonym: Succinic acid dimethyl ester

  • CAS Number 106-65-0

  • Linear Formula CH3OCOCH2CH2COOCH3

  • Molecular Weight 146.14

  •  Beilstein Registry Number 956776

  •  EC Number 203-419-9

  •  MDL number MFCD00008466

  •  PubChem Substance ID 24848500

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
vapor pressure   0.3 mmHg ( 20 °C)
InChI Key   MUXOBHXGJLMRAB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   689 °F
expl. lim.   8.5 %
refractive index   n20/D 1.419(lit.)
  n20/D 1.419
bp   200 °C(lit.)
mp   16-19 °C(lit.)
density   1.117 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
WM7675000
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?