EMAIL THIS PAGE TO A FRIEND
14260 Sigma-Aldrich

Beryllium oxide

purum

Purchase

Properties

Related Categories Beryllium, Catalysis and Inorganic Chemistry, Chemical Synthesis, Materials Science, Metal and Ceramic Science,
InChI Key   LTPBRCUWZOMYOC-UHFFFAOYSA-N
grade   purum
density   3.01 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1566 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

202770 Beryllium oxide, 99.98% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?