EMAIL THIS PAGE TO A FRIEND
14310 Sigma-Aldrich

BG11 Broth

for microbiology

Purchase

Properties

Related Categories A - M, All Liquid Media/Broths, Catalysis and Inorganic Chemistry, Chemical Synthesis, Cyanobacteria,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   calcium chloride dihydrate, 36.7 mg/L
  citric acid, 5.6 mg/L
  dipotassium hydrogen phosphate, 31.4 mg/L
  disodium magnesium EDTA, 1 mg/L
  ferric ammonium citrate, 6 mg/L
  magnesium sulfate, 36 mg/L
  sodium carbonate, 20 mg/L
  sodium nitrate, 1.5 g/L
suitability   Cyanobacteria

Description

Application

Universal medium for the culture and maintenance of cyanobacteria.

Other Notes

Suitable for photoautotrophic growth of Synechcystis sp. PCC 6803

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1498 5.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

73816 BG11 Broth, for microbiology, 100x concentrate, sterile filtered

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?