• USA Home
  • 14339 - Bicinchoninic acid disodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
14339 Sigma-Aldrich

Bicinchoninic acid disodium salt hydrate

≥99.0% (HPLC)

Synonym: 2,2′-Biquinoline-4,4′-dicarboxylic acid disodium salt, 4,4′-Dicarboxy-2,2′-biquinoline disodium salt, BCA, Disodium 2,2′-biquinoline-4,4′-dicarboxylate

Purchase

Properties

InChI Key   FAFVGCAYKZKOPT-UHFFFAOYSA-L
assay   ≥99.0% (HPLC)
impurities   ~12% water

Description

Other Notes

Photometric reagent for protein assay. Forms an intense purple complex with the cuprous ion and hence the basis of the analytical monitoring of cuprous ion produced in the biuret reaction. Demonstrates greater tolerance to interference from non-ionic detergents and simple buffer salts

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D8284 Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?