• USA Home
  • 14382 - (±)-1,1′-Bi(2-naphthol)

EMAIL THIS PAGE TO A FRIEND
14382 Sigma-Aldrich

(±)-1,1′-Bi(2-naphthol)

puriss., ≥99.0% (NT)

Synonym: (±)-1,1′-Binaphthalene-2,2′-diol, (±)-2,2′-Dihydroxy-1,1′-dinaphthyl

  • CAS Number 602-09-5

  • Empirical Formula (Hill Notation) C20H14O2

  • Molecular Weight 286.32

  •  Beilstein Registry Number 997518

  •  EC Number 210-014-0

  •  MDL number MFCD00004068

  •  PubChem Substance ID 24848612

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   PPTXVXKCQZKFBN-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
mp   215-218 °C
solubility   dioxane: soluble0.5 g/10 mL, clear, colorless to slightly yellow

Description

Other Notes

Zum Weinsäurenachweis

Same product — same quality — now Sigma-Aldrich brand

Application

(±)-1,1′-Bi(2-naphthol) was used to develop chiral fluorescent sensors for highly enantioselective detection of chiral amino alcohols, α-hydroxycarboxylic acids and amino acid derivatives. It was used in chiral separation by capillary electrophoresis using a combination of a polymerized chiral micelle and γ-cyclodextrin.

General description

(±)-1,1′-Bi(2-naphthol) forms chiral cocrystals with photochromic diarylethene having 2-methyl-4-pyridyl groups.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
DU3106000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?