• USA Home
  • 14402 - Biotin, immobilized on Dextran

EMAIL THIS PAGE TO A FRIEND
14402 Sigma-Aldrich

Biotin, immobilized on Dextran

≥99.0% (TLC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Detection, Fluorescent Probes, Labels, Particles and Stains More...
assay   ≥99.0% (TLC)
mol wt   Mr ~70000
extent of labeling   5-25 mol/mol substitution (biotin)
solubility   H2O: 50 mg/mL, slightly hazy, faintly yellow
storage temp.   2-8°C

Description

Application

Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components. Biotinylated dextran may be used to capture or detect avidin/strepavidin conjugated molecules.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?