• USA Home
  • 14481 - 1,3-Bis(ethylamino)propane (sym.)

EMAIL THIS PAGE TO A FRIEND
14481 Sigma-Aldrich

1,3-Bis(ethylamino)propane (sym.)

purum, ≥97.0% (GC)

Synonym: N,N′-Diethyl-1,3-propanediamine

  • CAS Number 10061-68-4

  • Linear Formula C2H5NH(CH2)3NHC2H5

  • Molecular Weight 130.23

  •  Beilstein Registry Number 1734025

  •  EC Number 233-200-3

  •  MDL number MFCD00009036

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   BEPGHZIEOVULBU-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.437
density   0.816 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

1,3-Bis(ethylamino)propane was used in the synthesis of complexes of Ru(II), Cu(II) and Mn(II) having an unsymmetrical macrocycle containing pyrazole and amine nitrogen atoms as donor sites. It was used in parallel batch screening technique for the identification of both selective and high-affinity displacers for a model binary mixture of proteins in a cation-exchange system.

General description

1,3-Bis(ethylamino)propane (N,N′-Diethyl-1,3-propanediamine) is a potential displacer of ribonuclease A and α-chymotrypsinogen A adsorbed together on high-performance sepharose.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN2 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
116.6 °F
Flash Point(C) 
47 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

138428 N,N′-Diethyl-1,3-propanediamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?