• USA Home
  • 14490 - Bis(2-ethylhexyl)amine

EMAIL THIS PAGE TO A FRIEND
14490 Sigma-Aldrich

Bis(2-ethylhexyl)amine

purum, ≥98.0% (GC)

Synonym: Diisooctylamine

  • CAS Number 106-20-7

  • Linear Formula [CH3(CH2)3CH(C2H5)CH2]2NH

  • Molecular Weight 241.46

  •  Beilstein Registry Number 1748342

  •  EC Number 203-372-4

  •  MDL number MFCD00009489

  •  PubChem Substance ID 329750507

Purchase

Properties

InChI Key   SAIKULLUBZKPDA-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.443(lit.)
  n20/D 1.443
bp   123 °C/5 mmHg(lit.)
density   0.805 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Bis(2-ethylhexyl)amine(BEA) was used in the synthesis of stable nanosized CdS particles coated by an oriented monolayer of chemically bonded BEA molecules.

General description

Bis(2-ethylhexyl)amine constitutes surfactant-based liquid mixtures having potential specialized applications.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
2
RTECS 
IH6825000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

292850 Bis(2-ethylhexyl)amine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?