• USA Home
  • 14503 - N,N′-Bis(2-aminoethyl)-1,3-propanediamine

EMAIL THIS PAGE TO A FRIEND
14503 Sigma-Aldrich

N,N′-Bis(2-aminoethyl)-1,3-propanediamine

purum, ≥97.0% (NT)

  • CAS Number 4741-99-5

  • Linear Formula H2NCH2CH2NH(CH2)3NHCH2CH2NH2

  • Molecular Weight 160.26

  •  Beilstein Registry Number 1742732

  •  EC Number 225-254-1

  •  MDL number MFCD00008174

  •  PubChem Substance ID 24848545

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   UWMHHZFHBCYGCV-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT)
refractive index   n20/D 1.494(lit.)
  n20/D 1.494
bp   142-145 °C/8 mmHg(lit.)
density   0.96 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

N,N′-Bis(2-aminoethyl)-1,3-propanediamine was used to study the regulation of gene that encodes the periplasmic superoxide dismutase of Escherichia coli in response to copper.

Biochem/physiol Actions

N,N′-Bis(2-aminoethyl)-1,3-propanediamine inhibits mitochondrial cytochrome c oxidase by causing cellular Cu deficiency in a human promyelocytic leukemia cell line (HL-60).

General description

N,N′-Bis(2-aminoethyl)-1,3-propanediamine is a high affinity Cu(II) chelator.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN2 8 / PGIII
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?