• USA Home
  • 14525 - 1,2-Bis(3-aminopropylamino)ethane

EMAIL THIS PAGE TO A FRIEND
14525 Sigma-Aldrich

1,2-Bis(3-aminopropylamino)ethane

purum

Synonym: 3,3′-Ethylenediiminodipropylamine

  • CAS Number 10563-26-5

  • Linear Formula NH2CH2CH2CH2NHCH2CH2NHCH2CH2CH2NH2

  • Molecular Weight 174.29

  •  Beilstein Registry Number 2203748

  •  EC Number 234-147-9

  •  MDL number MFCD00008210

  •  PubChem Substance ID 329750520

Purchase

Properties

InChI Key   RXFCIXRFAJRBSG-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (NT)
refractive index   n20/D 1.491(lit.)
  n20/D 1.492
bp   150-160 °C/5 mmHg(lit.)
density   0.952 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
RTECS 
TX8032950
Flash Point(F) 
287.6 °F
Flash Point(C) 
142 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?