• USA Home
  • 14801 - [(S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene]chloro(p-cymene)ruthenium chloride

EMAIL THIS PAGE TO A FRIEND
14801 Sigma-Aldrich

[(S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene]chloro(p-cymene)ruthenium chloride

purum

  • CAS Number 130004-33-0

  • Empirical Formula (Hill Notation) C54H46Cl2P2Ru

  • Molecular Weight 928.87

  •  MDL number MFCD00134456

Purchase

Properties

InChI Key   WNHLGYRPKARUHY-UHFFFAOYSA-L
grade   purum
optical activity   [α]20/D −570±10°, c = 0.1% in chloroform

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?