EMAIL THIS PAGE TO A FRIEND
15094 Sigma-Aldrich

Silver ionophore IV

Selectophore, function tested

Synonym: O,O″-Bis[2-(methylthio)ethyl]-tert-butylcalix[4]arene

  • CAS Number 145237-25-8

  • Empirical Formula (Hill Notation) C50H68O4S2

  • Molecular Weight 797.20

  •  Beilstein Registry Number 6170641

  •  MDL number MFCD00467948

  •  PubChem Substance ID 24849076

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   DKWOMOMMOUZRFI-UHFFFAOYSA-N
quality   function tested

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Ag-ionophore with excellent characteristics

Same product — same quality — now Sigma-Aldrich brand

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Silver ionophore IV was used for solvent extraction of silver trifluoromethanesulfonate from the two-phase water/nitrobenzene system.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

44042 Silver ionophore VII, Selectophore, function tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?