• USA Home
  • 15118 - Bis(4-nitrophenyl) carbonate

EMAIL THIS PAGE TO A FRIEND
15118 Sigma-Aldrich

Bis(4-nitrophenyl) carbonate

purum, ≥97.0% (CHN)

Synonym: 4-Nitrophenyl carbonate

  • CAS Number 5070-13-3

  • Linear Formula (O2NC6H4O)2CO

  • Molecular Weight 304.21

  •  Beilstein Registry Number 1892897

  •  EC Number 225-775-4

  •  MDL number MFCD00007322

  •  PubChem Substance ID 24849099

Purchase

Properties

Related Categories Active Esters/Additives, Chemical Synthesis, Coupling, Synthetic Reagents
InChI Key   ACBQROXDOHKANW-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (CHN)
mp   136-139 °C(lit.)
  137-142 °C

Description

Application

Reagent for the synthesis of 4-nitrophenyl active esters of amino acids. Used in the preparation of symmetrical and unsymmetrical ureas.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Coupling Reagents

In principle, the seemingly simple formation of a peptide bond can be accomplished using all the procedures available in organic chemistry for the synthesis of carboxylic acid amides. However, due to...
Matthias Junkers
Aldrich ChemFiles 2007, 7.2, 3.
Keywords: Acylations, Catalysis, Condensations, Dehydration reaction, Peptide synthesis, Racemizations, Solid phase peptide synthesis

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

161691 Bis(4-nitrophenyl) carbonate, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?