• USA Home
  • 15148 - (S,S)-(−)-N,N′-Bis(α-methylbenzyl)sulfamide

EMAIL THIS PAGE TO A FRIEND
15148 Sigma-Aldrich

(S,S)-(−)-N,N′-Bis(α-methylbenzyl)sulfamide

purum, ≥98.0% (sum of enantiomers, TLC)

Synonym: (S,S)-(−)-N,N′-Bis(1-phenylethyl)sulfamide

  • CAS Number 27304-75-2

  • Linear Formula [C6H5CH(CH3)NH]2SO2

  • Molecular Weight 304.41

  •  Beilstein Registry Number 4701487

  •  MDL number MFCD00064477

  •  PubChem Substance ID 329750745

Purchase

Properties

InChI Key   ZUMVQOILJDLACR-KBPBESRZSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, TLC)
optical activity   [α]20/D −78±2°, c = 2% in ethanol
mp   98-100 °C(lit.)
  98-100 °C

Description

Other Notes

Chiral modifier for asymmetric reductions

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

276073 (S,S)-(−)-N,N′-Bis(α-methylbenzyl)sulfamide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?