• USA Home
  • 15153 - Bis(phenylsulfonyl)methane

EMAIL THIS PAGE TO A FRIEND
15153 Sigma-Aldrich

Bis(phenylsulfonyl)methane

purum, ≥97.0% (HPLC)

Synonym: Methylenebis(phenyl sulfone)

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfones, Sulfur Compounds More...
InChI Key   QCHNSJNRFSOCLJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
mp   119-121 °C(lit.)
  119-121 °C

Description

Other Notes

Activated C1-building block; it can be dialkylated and desulfonylated (methylene-dianion equivalent) ; Pd-catalyzed alkylation of the anion

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?