• USA Home
  • 15190 - N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide

EMAIL THIS PAGE TO A FRIEND
15190 Sigma-Aldrich

N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide

technical, ≥95% (HPLC)

Synonym: N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulfonamide, N,O,O′-Tritosyldiethanolamine

  • CAS Number 16695-22-0

  • Linear Formula CH3C6H4SO2N(CH2CH2OSO2C6H4CH3)2

  • Molecular Weight 567.69

  •  Beilstein Registry Number 2406580

  •  EC Number 412-920-3

  •  MDL number MFCD00026000

  •  PubChem Substance ID 329750767

Purchase

Properties

InChI Key   VJDZYMIEDBLSDT-UHFFFAOYSA-N
grade   technical
assay   ≥95% (HPLC)
mp   92-98 °C(lit.)
  92-98 °C

Description

Other Notes

Building block for the synthesis of various aza-oxa-thia macrocyclic ligands

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

380687 N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?