EMAIL THIS PAGE TO A FRIEND
15215 Sigma-Aldrich

Benzoyl chloride

purum, ≥99% (GC)

Purchase

Properties

vapor density   4.88 (vs air)
vapor pressure   1 mmHg ( 32 °C)
InChI Key   PASDCCFISLVPSO-UHFFFAOYSA-N
grade   purum
assay   ≥99% (GC)
autoignition temp.   1056 °F
expl. lim.   4.9 %
refractive index   n20/D 1.553(lit.)
bp   198 °C(lit.)
mp   −1 °C(lit.)
density   1.211 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1736 8 / PGII
WGK Germany 
1
RTECS 
DM6600000
Flash Point(F) 
161.6 °F
Flash Point(C) 
72 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?