• USA Home
  • 15231 - Bis(trimethylsilyl)carbodiimide

EMAIL THIS PAGE TO A FRIEND
15231 Sigma-Aldrich

Bis(trimethylsilyl)carbodiimide

purum, ≥97.0% (GC)

Synonym: 1,3-Bis(trimethylsilyl)carbodiimide, N,N′-Methanetetraylbis[1,1,1-trimethylsilylamine], N,N-Bis(trimethylsilyl)carbodiimide

  • CAS Number 1000-70-0

  • Linear Formula (CH3)3SiN=C=NSi(CH3)3

  • Molecular Weight 186.40

  •  Beilstein Registry Number 2042082

  •  EC Number 213-673-2

  •  MDL number MFCD00051538

Purchase

Properties

Related Categories C-X Bond Formation (Non-Halogen), Chemical Synthesis, Other Non-Halogen Reagents for C-X Bond Formation, Synthetic Reagents
InChI Key   KSVMTHKYDGMXFJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.434(lit.)
  n20/D 1.434
bp   164 °C(lit.)
mp   −23 °C(lit.)
density   0.821 g/mL at 25 °C(lit.)

Description

Other Notes

Highly reactive synthetic equivalent of cyanamide: synthesis of N,N′-dicyano-quinodiimines from quinones

Same product — same quality — now Sigma-Aldrich brand

Application

Reactant for preparation of:
• 4-substituted methoxylbenzoyl-aryl-thiazole analogues as potent and orally bioavailable anticancer agents
• O-silylurethanes
• Porous SiCN materials
• Luminescent diketo-pyrrolo-pyrrole analog
• Hard thin films of silicon carbonitride SiCN as potential protection coatings against wear and corrosion of metals
• Ceramic materials

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
3
RTECS 
VV1302500
Flash Point(F) 
69.8 °F
Flash Point(C) 
21 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

344338 Bis(trimethylsilyl)carbodiimide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?