• USA Home
  • 15233 - N,O-Bis(trimethylsilyl)hydroxylamine

EMAIL THIS PAGE TO A FRIEND
15233 Sigma-Aldrich

N,O-Bis(trimethylsilyl)hydroxylamine

purum, ≥97.0% (GC)

  • CAS Number 22737-37-7

  • Linear Formula (CH3)3SiONHSi(CH3)3

  • Molecular Weight 177.39

  •  Beilstein Registry Number 1920452

  •  EC Number 245-188-7

  •  MDL number MFCD00008266

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydroxylamines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   ZAEUMMRLGAMWKE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.411(lit.)
bp   78-80 °C/100 mmHg(lit.)
density   0.83 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Reagent for the synthesis of O-silylated hydroxamates which can be transformed into isocyanates ; Synthesis of oximes from carbonyl compounds; Reagent for electrophilic amination; Reagent for nucleophilic hydroxylamination

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
84.2 °F
Flash Point(C) 
29 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

235105 N,O-Bis(trimethylsilyl)hydroxylamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?