• USA Home
  • 15242 - N,O-Bis(trimethylsilyl)acetamide

EMAIL THIS PAGE TO A FRIEND
15242 Sigma-Aldrich

N,O-Bis(trimethylsilyl)acetamide

purum, ≥95.0% (GC)

Synonym: BSA

  • CAS Number 10416-59-8

  • Linear Formula CH3C[=NSi(CH3)3]OSi(CH3)3

  • Molecular Weight 203.43

  •  Beilstein Registry Number 1306669

  •  EC Number 233-892-7

  •  MDL number MFCD00008270

  •  PubChem Substance ID 329750797

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents GC, Reagents for Silylation,
InChI Key   SIOVKLKJSOKLIF-CMDGGOBGSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.417(lit.)
  n20/D 1.418
bp   71-73 °C/35 mmHg(lit.)
density   0.832 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
RTECS 
AK3000000
Flash Point(F) 
111.2 °F
Flash Point(C) 
44 °C
Protocols & Articles

Articles

Silylating Reagents for GC-MS Compatible Derivatives of NSAIDs

Human and veterinary pharmaceuticals are a group of “emerging” contaminants,1, 2 some of which are produced in increasingly large volumes every year. The amounts produced are reaching quantities simi...
Luis-Alberto Martin
AnalytiX Volume 10 Article 2
Keywords: Anti-inflammatory agents, Derivatizations, Environmental, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Metabolites, Pesticides, Pharmaceutical, Silylations, Veterinary

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

128910 N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?