• USA Home
  • 15266 - Bis(trimethylsilyl) phosphite

EMAIL THIS PAGE TO A FRIEND
15266 Sigma-Aldrich

Bis(trimethylsilyl) phosphite

purum, ≥98.0% (GC)

  • CAS Number 3663-52-3

  • Empirical Formula (Hill Notation) C6H19O3PSi2

  • Molecular Weight 226.36

  •  Beilstein Registry Number 1776928

  •  MDL number MFCD00210018

  •  PubChem Substance ID 329750806

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Organic Phosphates/Phosphites, Phosphorus Compounds More...
InChI Key   TVYLOQJEHUAACN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.412
bp   77-79 °C/11 mmHg
density   0.968 g/mL at 20 °C(lit.)

Description

Other Notes

Reagent for the preparation of α-amino phosphonic acids by addition to imines; Preparation of phosphoric acid derivatives; Synthesis of phosphonoformates, interesting antivirals

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
136.4 °F
Flash Point(C) 
58 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?