• USA Home
  • 15277 - 3,5-Bis(trifluoromethyl)benzylamine

EMAIL THIS PAGE TO A FRIEND
15277 Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzylamine

technical, ~80% (GC)

  • CAS Number 85068-29-7

  • Linear Formula (CF3)2C6H3CH2NH2

  • Molecular Weight 243.15

  •  Beilstein Registry Number 7209267

  •  EC Number 285-290-9

  •  MDL number MFCD00009909

Purchase

Properties

InChI Key   DHVHORCFFOSRBP-UHFFFAOYSA-N
grade   technical
assay   ≥90% (NT)
  ~80% (GC)
mp   50-55 °C(lit.)
  50-55 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3259 8/PG 2
WGK Germany 
3
Flash Point(F) 
173 °F
Flash Point(C) 
78 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

263389 3,5-Bis(trifluoromethyl)benzylamine, technical grade, 80%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?