• USA Home
  • 15293 - Blotting-Nylon 66 membranes, type B, positive

EMAIL THIS PAGE TO A FRIEND
15293 Sigma-Aldrich

Blotting-Nylon 66 membranes, type B, positive

sheet, pkg of 10 membranes

Synonym: Nylon 66-Blotting membranes, type B

Purchase

Properties

Related Categories Membranes & Blotting Paper, Molecular Biology, Nylon 66 Membranes, Positively Charged, Protein Electrophoresis, Proteomics,
grade   for molecular biology
form   sheet
packaging   pkg of 10 membranes
size   215 mm × 215 mm
surface coverage   surface (quaternary ammonium groups) 100%
pore size   0.45 μm pore size

Description

General description

Positively charged nylon membrane, strengthened with polyester; high tenside strength and breaking strength

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?