EMAIL THIS PAGE TO A FRIEND
15294 Sigma-Aldrich

Boc-GABA-OH

purum, ≥99.0% (T)

Synonym: 4-(Boc-amino)butyric acid, Boc-GABA

  • CAS Number 57294-38-9

  • Empirical Formula (Hill Notation) C9H17NO4

  • Molecular Weight 203.24

  •  Beilstein Registry Number 1869578

  •  MDL number MFCD00037313

  •  PubChem Substance ID 329750811

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Linear Core Amino Acids, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   HIDJWBGOQFTDLU-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   55-59 °C
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?