EMAIL THIS PAGE TO A FRIEND
15334 Sigma-Aldrich

Lead(II) nitrate

puriss. p.a., ACS reagent, ≥99.0% (T)

Synonym: Lead dinitrate

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, E-L, Puriss p.a. ACS, Essential Chemicals,
grade   ACS reagent
InChI Key   RLJMLMKIBZAXJO-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99.0% (T)
form   solid
mp   470 °C (dec.)(lit.)
anion traces   chloride (Cl-): ≤5 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Application

Several new polymeric complexes of lead with dialkylamino derivatives of squaric acid were produced to study their coordination modes as possible prototypes for studies of lanthanide complexes.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?