EMAIL THIS PAGE TO A FRIEND
15426 Sigma-Aldrich

Boc-His-OH

puriss., ≥99.0% (T)

Synonym: Nα-(tert-Butoxycarbonyl)-L-histidine, Nα-Boc-L-histidine

  • CAS Number 17791-52-5

  • Empirical Formula (Hill Notation) C11H17N3O4

  • Molecular Weight 255.27

  •  Beilstein Registry Number 755289

  •  EC Number 241-768-9

  •  MDL number MFCD00065576

  •  PubChem Substance ID 329750874

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Histidine, Peptide Chemistry More...
InChI Key   AYMLQYFMYHISQO-QMMMGPOBSA-N
grade   puriss.
assay   ≥99.0% (T)
optical activity   [α]20/D +26±1°, c = 1% in methanol
mp   195 °C (dec.)(lit.)
  ~195 °C (dec.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

408433 Boc-His-OH, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?