EMAIL THIS PAGE TO A FRIEND
154687 Sigma-Aldrich

2-Methyl-1-propanol

ACS spectrophotometric grade, ≥99.0%

Synonym: Isobutanol, Isobutyl alcohol

Purchase

Properties

Related Categories Amber Glass Bottles, Solvent Bottles, Solvent Packaging Options, Solvent by Application, Solvents,
grade   ACS spectrophotometric grade
vapor density   2.55 (vs air)
vapor pressure   8 mmHg ( 20 °C)
  8.8 mmHg ( 0 °C)
InChI Key   ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
assay   ≥99.0%
autoignition temp.   801 °F
expl. lim.   10.6 %
impurities   ≤0.0005 meq/g Titr. acid
  ≤0.01% butyraldehyde
  ≤0.02% 2-butanone
  ≤0.1% water
evapn. residue   <0.001%
color   APHA: ≤10
refractive index   n20/D 1.396(lit.)
bp   108 °C(lit.)
mp   −108 °C(lit.)
density   0.803 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 230 nm Amax: 1.00
  λ: 240 nm Amax: 0.25
  λ: 250 nm Amax: 0.07
  λ: 260 nm Amax: 0.05
  λ: 330-400 nm Amax: 0.01

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.





Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1212 - class 3 - PG 3 - Isobutanol
WGK Germany 
1
RTECS 
NP9625000
Flash Point(F) 
82.4 °F
Flash Point(C) 
28 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?