EMAIL THIS PAGE TO A FRIEND
154733 Sigma-Aldrich

Chloroform

ACS spectrophotometric grade, ≥99.8%, contains amylenes as stabilizer

Synonym: Methylidyne trichloride, Trichloromethane

Purchase

Properties

Related Categories Amber Glass Bottles, Chloroform, Solvent by Type, Solvent Bottles, Solvent Packaging Options,
grade   ACS spectrophotometric grade
vapor density   4.1 (vs air)
vapor pressure   160 mmHg ( 20 °C)
InChI Key   HEDRZPFGACZZDS-UHFFFAOYSA-N
assay   ≥99.8%
contains   amylenes as stabilizer
packaging   glass bottle of 2 L
impurities   Acetone and aldehyde, passes test
  Acid and chloride, passes test
  Free chlorine (Cl), passes test
  ≤0.03% water
evapn. residue   <0.001%
color   APHA: ≤10
refractive index   n20/D 1.445(lit.)
bp   60.5-61.5 °C(lit.)
mp   −63 °C(lit.)
density   1.492 g/mL at 25 °C(lit.)
  1.492 g/mL at 25 °C
cation traces   Pb: ≤0.05 ppm
λ   H2O reference
UV absorption   λ: 245 nm Amax: 1.00
  λ: 255 nm Amax: 0.25
  λ: 260 nm Amax: 0.15
  λ: 270 nm Amax: 0.05
  λ: 290-400 nm Amax: 0.01

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

2 L in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1888 6.1 / PGIII
WGK Germany 
3
RTECS 
FS9100000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?