154903 Sigma-Aldrich

Methanol

ACS spectrophotometric grade, ≥99.9%

DOWNLOAD MSDS (PDF)

Synonym: Methyl alcohol

Properties

Related Categories Amber Glass Bottles, Methanol, Solvent by Type, Solvent Bottles, Solvent Packaging Options,
grade   ACS spectrophotometric grade
vapor density   1.11 (vs air)
vapor pressure   410 mmHg ( 50 °C)
  97.68 mmHg ( 20 °C)
assay   ≥99.9%
autoignition temp.   725 °F
expl. lim.   36 %
impurities   MnO4- reducers, passes test
  ≤0.0002 meq/g Titr. base
  ≤0.0003 meq/g Titr. acid
  ≤0.001% acetaldehyde
  ≤0.001% acetone
  ≤0.001% formaldehyde
  ≤0.1% water
evapn. residue   ≤0.001%
color   APHA: ≤10
  clear
refractive index   n20/D 1.329(lit.)
bp   64.7 °C(lit.)
mp   −98 °C(lit.)
density   0.791 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 205 nm Amax: 1.00
  λ: 210 nm Amax: 0.80
  λ: 220 nm Amax: 0.40
  λ: 230 nm Amax: 0.20
  λ: 240 nm Amax: 0.10
  λ: 260 nm Amax: 0.04
  λ: 280-400 nm Amax: 0.01

Description

Packaging

1, 6×1, 2, 4×2, 4×4 L in glass bottle

500 mL in glass bottle

Preparation Note

Product filtered through a 0.2 μm filter

Price and Availability

Customers Also Viewed

Acetone
154598Acetone

ACS spectrophotometric grade, ≥99.5%

Methanol

CHROMASOLV®, for HPLC, ≥99.9%

Methanol
322415Methanol

anhydrous, 99.8%

Methanol

spectrophotometric grade, ≥99%

Methanol
179337Methanol

ACS reagent, ≥99.8%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?