• USA Home
  • 15622 - Hydrazine hydrate solution

EMAIL THIS PAGE TO A FRIEND
15622 Sigma-Aldrich

Hydrazine hydrate solution

24-26% (iodometric)

Synonym: Hydrazinium hydroxide

  • CAS Number 10217-52-4

  • Linear Formula NH2NH2 · H2O

  • Molecular Weight 50.06

  •  Beilstein Registry Number 878137

  •  MDL number MFCD00149931

  •  PubChem Substance ID 24849598

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
assay   24-26% (iodometric)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 3293 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles

Articles

Determination of Water Content in Hydrazine hydrate solution Using Karl Fischer Titration

Under standard conditions Hydrazine Hydrate solution causes a side reaction with the iodine present in a KF-titration. The side-reaction can be suppressed if the working medium in the titration vesse...
Keywords: Titrations

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?