EMAIL THIS PAGE TO A FRIEND
15680 Sigma-Aldrich

Boric anhydride

purum p.a., ≥97.0% (T)

Synonym: Boron trioxide

  • CAS Number 1303-86-2

  • Empirical Formula (Hill Notation) B2O3

  • Molecular Weight 69.62

  •  EC Number 215-125-8

  •  MDL number MFCD00011315

  •  eCl@ss 38120103

Purchase

Properties

vapor density   >1 (vs air)
InChI Key   JKWMSGQKBLHBQQ-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥97.0% (T)
loss   ≤3% loss on ignition, 900 °C
mp   450 °C(lit.)
density   2.46 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤2000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?