• USA Home
  • 15692 - Boron tribromide solution

EMAIL THIS PAGE TO A FRIEND
15692 Sigma-Aldrich

Boron tribromide solution

purum, ~1 M in methylene chloride

Synonym: Tribromoboron

  • CAS Number 10294-33-4

  • Empirical Formula (Hill Notation) BBr3

  • Molecular Weight 250.52

  •  MDL number MFCD00011312

Purchase

Properties

Related Categories Chemical Synthesis, Deprotecting Reagents, Other Deprotecting Reagents, Protection and Derivatization, Synthetic Reagents More...
InChI Key   ILAHWRKJUDSMFH-UHFFFAOYSA-N
grade   purum
concentration   ~1 M in methylene chloride
density   1.46 g/mL at 20 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Reactant for preparation of:
• Drug intermediate 6-nitro-L-DOPA
• Luminescent polystyrene derivatives with sterically protected carbazolylborane moieties
• High-quality boron-doped graphene via Wurtz-type reductive coupling reaction
• Mercapto-(+)-methamphetamine haptens for synthesis of (+)-methamphetamine conjugate vaccines with improved epitope densities
• Micrometer-sized organic molecule-DNA hybrid structures
• Borane complexes via electrophilic aromatic borylation reactions
• A 5-HT2C receptor agonist
• Biphenyl-derivatives possessing tertiary amino groups as ß-secretase(BACE1) inhibitors for the treatment of Alzheimer′s disease
• A highly near-IR region fluorescent p-extended boron aza-dipyrromethene moiety unit
• Tetrahydroisoquinoline derivatives via intramolecular cyclization of methoxy-substituted N-phenethylimides

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Target organs 
Blood, Central nervous system, Liver
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3390 8(6.1) / PGI
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

211222 Boron tribromide solution, 1.0 M in methylene chloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?