• USA Home
  • 15715 - Boron trifluoride-methanol solution

EMAIL THIS PAGE TO A FRIEND
15715 Sigma-Aldrich

Boron trifluoride-methanol solution

~10% (~1.3 M), for synthesis

Purchase

Properties

Related Categories Acids, Acids & Bases, Boron, Catalysis and Inorganic Chemistry, Chemical Synthesis,
grade   for synthesis
InChI Key   JBXYCUKPDAAYAS-UHFFFAOYSA-N
concentration   ~10% (~1.3 M)
density   0.859 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 3286 8(6.1)(3) / PGII
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?