Synonym: Hydroxylammonium chloride
CAS Number 5470-11-1
Linear Formula NH2OH · HCl
Molecular Weight 69.49
Beilstein Registry Number 3539763
EC Number 226-798-2
MDL number MFCD00051089
PubChem Substance ID 24849814
Popular Documents: Specification Sheet (PDF)
| Related Categories | Bioactive Small Molecules, Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Enzyme Inhibitors, |
| Quality Level | PREMIUM |
| grade | ReagentPlus® |
| assay | 99% |
| mp | 155-157 °C (dec.)(lit.) |
| density | 1.67 g/mL at 25 °C(lit.) |
MAO inhibitor; inhibits platelet aggregation.
100, 500 g in poly bottle
3 kg in poly drum
ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?