EMAIL THIS PAGE TO A FRIEND
159697 Sigma-Aldrich

4-Nitroazobenzene

technical grade, 90%

Synonym: NAB

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, N, Stains & Dyes, A to Z, Stains and Dyes More...
grade   technical grade
InChI Key   TZTDJBMGPQLSLI-BUHFOSPRSA-N
assay   90%
mp   132-134 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CN2710000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?