EMAIL THIS PAGE TO A FRIEND
16000 Sigma-Aldrich

Pyruvonitrile

technical, ≥90% (GC)

Synonym: 2-Oxopropionitrile, Acetyl cyanide

  • CAS Number 631-57-2

  • Linear Formula CH3COCN

  • Molecular Weight 69.06

  •  Beilstein Registry Number 1737633

  •  MDL number MFCD00001838

  •  PubChem Substance ID 24849842

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   QLDHWVVRQCGZLE-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
refractive index   n20/D 1.376(lit.)
  n20/D 1.376
bp   92-93 °C(lit.)
density   0.974 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3273 6.1(3) / PGII
WGK Germany 
3
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

269212 Pyruvonitrile, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?