• USA Home
  • 16002 - SP Brij® C2 MBAL-SO-(SG)

EMAIL THIS PAGE TO A FRIEND
16002 Sigma-Aldrich

SP Brij® C2 MBAL-SO-(SG)

main component: diethylene glycol hexadecyl ether

Synonym: Brij® 52, Polyethylene glycol hexadecyl ether, Polyoxyethylene (2) cetyl ether

  • CAS Number 9004-95-9

  • Linear Formula C16H33(OCH2CH2)nOH, n~2

  •  Beilstein Registry Number 1867431

  •  PubChem Substance ID 24849845

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Materials Science, Micro/NanoElectronics,
vapor pressure   0.26 psi ( 20 °C)
description   non-ionic
refractive index   n20/D 1.466(lit.)
mp   36-38 °C
HLB value   5.3
density   0.978 g/mL at 25 °C(lit.)

Description

Legal Information

Brij is a registered trademark of Croda International PLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
TR1581470
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?