• USA Home
  • 16026 - Brilliant Green Phenol Red Lactose Sucrose Agar

EMAIL THIS PAGE TO A FRIEND
16026 Sigma-Aldrich

Brilliant Green Phenol Red Lactose Sucrose Agar

for microbiology

Purchase

Properties

Related Categories A - M, All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Clinical Media,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 13 g/L
  brilliant green, 0.0125 g/L
  lactose, 10 g/L
  mixed peptone, 10 g/L
  phenol red, 0.08 g/L
  sodium chloride, 5 g/L
  sucrose, 10 g/L
  yeast extract, 3 g/L
final pH   6.9±0.2 (25 °C)
suitability   selective and differential for Salmonella

Description

Application

Selective medium for the isolation of salmonellae, with the exception of S. typhi, from pathological material, stool, urine, food etc.

Other Notes

Isolation of salmonellae from environmental samples

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Differentiation and Identification Media for Salmonella

Salmonella contamination is the second leading cause of food-borne illness worldwide. Controlling outbreaks of Salmonella is an important task for food regulators, restaurants and the food industry i...
Jvo Siegrist
Microbiology Focus Edition 1.3
Keywords: Biochemistry, Fermentation, Food & Beverage, Indicators, Microbiology

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?